Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293641.1 | NP_001280570.1 | 8, 9 | 804 | ||
NM_146141.2 | NP_666253.1 | 8, 9 | 804 | |||
GenBank mRNA | AK003660.1 | - | 3, 4 | 1089 | ||
AK090384.1 | - | 8, 9 | 793 | |||
AK143475.1 | - | 8, 9 | 775 | |||
BC011417.1 | - | 8, 9 | 798 | 2654305 |