Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183109.3 | NP_898932.2 | 3 | 498 | ||
GenBank mRNA | AK015583.1 | - | 3 | 510 | ||
AK132967.1 | - | 3 | 499 |