Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175217.6 | NP_780426.1 | 4 | 477 | ||
GenBank mRNA | AK132153.1 | - | 526 | |||
AY134664.1 | - | 4 | 418 | |||
AY424299.1 | - | 4 | 477 | |||
BC025064.1 | - | 4 | 477 | 4502289 |