Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113367.1 | NP_001106838.1 | 2 | 352 | ||
NM_029267.3 | NP_083543.2 | 2 | 297 | |||
GenBank mRNA | AF272859.1 | - | 2 | 160 | ||
AK016125.1 | - | 2 | 298 | |||
AK016403.1 | - | 2 | 352 | |||
CJ046602.1 | - | 2 | 352 |