Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnali1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175223.4 | NP_780432.1 | 2 | 254 | ||
GenBank mRNA | AK006256.1 | - | 231 | |||
AK077109.1 | - | 2 | 255 | |||
BC118619.1 | - | 2 | 229 | 40107304 | ||
BC122552.1 | - | 2 | 229 | 40107303 |