Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102468.1 | NP_001095938.1 | 4 | 358 | ||
NM_138304.2 | NP_612177.1 | 5 | 499 | |||
GenBank mRNA | AK008039.1 | - | 5 | 485 | ||
AY061807.1 | - | 5 | 499 | |||
BC013541.1 | - | 5 | 477 | 4241711 | ||
BC023475.1 | - | 5 | 464 | 5042406 | ||
BY097052.1 | - | 4 | 367 | |||
CO039047.1 | - | 1 | 374 |