Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029530.2 | NP_083806.2 | 3 | 740 | ||
GenBank mRNA | AK014127.1 | - | 1 | 182 | ||
AK018222.1 | - | 3 | 439 | |||
AK045946.1 | - | 2 | 792 | |||
BC004791.1 | - | 1 | 16 | 3588577 |