Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 5 | 490 | ||
NM_001165992.1 | NP_001159464.1 | 5 | 471 | |||
NM_001165995.1 | NP_001159467.1 | 4 | 377 | |||
NM_029702.4 | NP_083978.3 | 5 | 484 | |||
GenBank mRNA | AJ413953.1 | - | 5 | 437 | ||
AK005174.1 | - | 5 | 432 | |||
AK044759.1 | - | 5 | 442 | |||
AK050136.1 | - | 2 | 1342 | |||
AK083490.1 | - | 1 | 2867 | |||
AK180346.1 | - | 2 | 89 | |||
AK185500.1 | - | 2 | 90 | |||
BC010713.1 | - | 5 | 431 | 2649462 | ||
BC046782.1 | - | 5 | 422 | 5720540 | ||
BC046786.1 | - | 5 | 422 | 6416321 |