Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 7, 8 | 735 | ||
NM_001165992.1 | NP_001159464.1 | 7, 8 | 716 | |||
NM_001165995.1 | NP_001159467.1 | 6, 7 | 622 | |||
NM_029702.4 | NP_083978.3 | 7, 8 | 729 | |||
GenBank mRNA | AJ413953.1 | - | 7, 8 | 682 | ||
AK005174.1 | - | 7, 8 | 677 | |||
AK032592.1 | - | 1, 2 | 513 | |||
AK044759.1 | - | 7, 8 | 687 | |||
AK050136.1 | - | 4, 5 | 1587 | |||
BC010713.1 | - | 7, 8 | 676 | 2649462 | ||
BC046782.1 | - | 6, 7 | 596 | 5720540 | ||
BC046786.1 | - | 6, 7 | 596 | 6416321 |