Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146119.1 | NP_001139591.1 | 6, 7 | 675 | ||
NM_029758.4 | NP_084034.2 | 7, 8 | 748 | |||
GenBank mRNA | AK010821.1 | - | 5, 6 | 611 | ||
AK047142.1 | - | 6, 7 | 638 | |||
BC049664.1 | - | 7, 8 | 710 | 6532813 | ||
BC052465.1 | - | 6, 7 | 656 | 6834768 |