Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256112.1 | NP_001243041.1 | 5 | 568 | ||
NM_029759.4 | NP_084035.1 | 5 | 519 | |||
GenBank mRNA | AK010825.1 | - | 5 | 560 | ||
AK080126.1 | - | 4 | 2888 | |||
AK145362.1 | - | 5 | 560 | |||
AK146416.1 | - | 4 | 395 | |||
AK164004.1 | - | 4 | 395 | |||
AK169047.1 | - | 5 | 427 | |||
AK171703.1 | - | 5 | 560 | |||
AK216840.1 | - | 4 | 385 | |||
BC026808.1 | - | 4 | 391 | 4159289 |