Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256112.1 | NP_001243041.1 | 3, 4 | 418 | ||
NM_029759.4 | NP_084035.1 | 3, 4 | 369 | |||
GenBank mRNA | AK010825.1 | - | 3, 4 | 410 | ||
AK080126.1 | - | 2, 3 | 2738 | |||
AK089141.1 | - | 3, 4 | 410 | |||
AK145362.1 | - | 3, 4 | 410 | |||
AK146416.1 | - | 2, 3 | 245 | |||
AK164004.1 | - | 2, 3 | 245 | |||
AK169047.1 | - | 3, 4 | 277 | |||
AK171703.1 | - | 3, 4 | 410 | |||
AK187934.1 | - | 3, 4 | 250 | |||
AK216840.1 | - | 2, 3 | 235 | |||
BC026808.1 | - | 2, 3 | 241 | 4159289 |