Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133771.2 | NP_598532.1 | 7 | 741 | ||
GenBank mRNA | AF363447.1 | - | 8 | 803 | ||
AK002639.1 | - | 2 | 146 | |||
AK159560.1 | - | 7 | 720 | |||
BC013819.1 | - | 7 | 758 | 3489363 |