Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 15 | 1592 | ||
NM_001205392.1 | NP_001192321.1 | 13 | 1487 | |||
NM_029846.4 | NP_084122.2 | 14 | 1544 | |||
GenBank mRNA | AB087879.1 | - | 13 | 1298 | ||
AB087880.1 | - | 14 | 1355 | |||
AB087881.1 | - | 15 | 1403 | |||
AB476646.1 | - | 15 | 1403 | |||
AK005181.1 | - | 14 | 1761 | |||
AK030983.1 | - | 14 | 1485 | |||
AK131120.1 | - | 12 | 2574 | |||
AK155358.1 | - | 13 | 1423 | |||
AK170073.1 | - | 13 | 1421 | |||
BC033362.1 | - | 4 | 1109 | |||
BC049122.1 | - | 15 | 1517 | 6813377 | ||
BC052087.1 | - | 2 | 442 |