Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 9 | 1446 | ||
NM_029898.3 | NP_084174.2 | 8 | 1340 | |||
GenBank mRNA | AK045480.1 | - | 8 | 974 | ||
AK138273.1 | - | 8 | 995 | |||
AK140417.1 | - | 9 | 1310 | |||
BC152317.1 | - | 8 | 967 | 40088531 | ||
BC152536.1 | - | 8 | 967 | 40088530 |