Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 10 | 2105 | ||
NM_029898.3 | NP_084174.2 | 9 | 1999 | |||
GenBank mRNA | AK045480.1 | - | 9 | 1633 | ||
AK140417.1 | - | 10 | 1969 | |||
BC152317.1 | - | 9 | 1626 | 40088531 | ||
BC152536.1 | - | 9 | 1626 | 40088530 |