Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029963.2 | NP_084239.1 | 1, 2 | 66 | ||
GenBank mRNA | AB049941.1 | - | 1, 2 | 66 | ||
AK018829.1 | - | 1, 2 | 74 | |||
AK078198.1 | - | 1, 2 | 74 | |||
AK083366.1 | - | 1, 2 | 68 | |||
AK155101.1 | - | 1, 2 | 82 | |||
BC021947.1 | - | 1, 2 | 64 | 5098933 |