Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029963.2 | NP_084239.1 | 11 | 993 | ||
GenBank mRNA | AB049941.1 | - | 11 | 993 | ||
AK078198.1 | - | 10 | 2252 | |||
AK083366.1 | - | 11 | 995 | |||
AK155101.1 | - | 11 | 1009 | |||
AK183617.1 | - | 2 | 76 | |||
BC021947.1 | - | 11 | 991 | 5098933 |