Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 16 | 1616 | ||
NM_001159966.1 | NP_001153438.1 | 16 | 1514 | |||
NM_001159967.1 | NP_001153439.1 | 15 | 1538 | |||
NM_001159968.1 | NP_001153440.1 | 15 | 1511 | |||
NM_023884.4 | NP_076373.3 | 16 | 1556 | |||
GenBank mRNA | AF312924.1 | - | 15 | 1464 | ||
AK018622.1 | - | 16 | 1558 | |||
AK019543.1 | - | 15 | 1514 | |||
AK029049.1 | - | 16 | 1557 | |||
AK029993.1 | - | 16 | 1616 | |||
AK033549.1 | - | 16 | 1512 | |||
AK036803.1 | - | 15 | 1488 | |||
AK161080.1 | - | 15 | 1505 | |||
AK165002.1 | - | 16 | 1508 | |||
AK169368.1 | - | 15 | 1456 | |||
BC043132.1 | - | 15 | 1443 | 6811441 | ||
BC052663.1 | - | 16 | 1533 | 30051209 |