Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 18, 19 | 1866 | ||
NM_001159966.1 | NP_001153438.1 | 18, 19 | 1764 | |||
NM_001159967.1 | NP_001153439.1 | 17, 18 | 1788 | |||
NM_001159968.1 | NP_001153440.1 | 17, 18 | 1761 | |||
NM_023884.4 | NP_076373.3 | 18, 19 | 1806 | |||
GenBank mRNA | AF312924.1 | - | 17, 18 | 1714 | ||
AK007570.1 | - | 1, 2 | 126 | |||
AK019543.1 | - | 17, 18 | 1764 | |||
AK029993.1 | - | 18, 19 | 1866 | |||
AK033549.1 | - | 18, 19 | 1762 | |||
AK036803.1 | - | 17, 18 | 1738 | |||
AK169368.1 | - | 17, 18 | 1706 | |||
BC043132.1 | - | 17, 18 | 1693 | 6811441 | ||
BC052663.1 | - | 18, 19 | 1783 | 30051209 |