Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024467.3 | NP_077787.3 | 2 | 1139 | ||
GenBank mRNA | AK048624.1 | - | 2 | 1000 | ||
AK052274.1 | - | 2 | 974 | |||
AK089263.1 | - | 2 | 990 | |||
AK122500.1 | - | 297 | ||||
BC033441.1 | - | 2 | 965 | |||
BC059823.1 | - | 2 | 1139 | 6843792 |