Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 2 | 519 | ||
AK002290.1 | - | 2 | 413 | |||
AK146657.1 | - | 2 | 481 | |||
AK158530.1 | - | 2 | 378 | |||
AK160924.1 | - | 2 | 415 | |||
AK164392.1 | - | 2 | 444 | |||
AK167718.1 | - | 2 | 418 | |||
AK168220.1 | - | 2 | 432 | |||
AK168291.1 | - | 2 | 432 | |||
AK187532.1 | - | 2 | 222 | |||
AK200247.1 | - | 2 | 224 | |||
BC012962.1 | - | 2 | 417 | 4188579 |