Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 1 | 268 | ||
AK002290.1 | - | 1 | 163 | |||
AK146657.1 | - | 1 | 230 | |||
AK160924.1 | - | 1 | 164 | |||
AK164392.1 | - | 1 | 193 | |||
AK167718.1 | - | 1 | 167 | |||
AK168220.1 | - | 1 | 189 | |||
AK168291.1 | - | 1 | 189 | |||
BC012962.1 | - | 1 | 166 | 4188579 | ||
BY251864.1 | - | 1 | 286 |