Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145501.2 | NP_663476.1 | 5 | 1006 | ||
GenBank mRNA | AK030725.1 | - | 5 | 971 | ||
BC089309.1 | - | 5 | 947 | |||
BC110363.1 | - | 5 | 968 | 5346917 |