Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033146.1 | NP_149158.1 | 3, 4 | 486 | ||
GenBank mRNA | AB054001.1 | - | 3, 4 | 532 | ||
AK005154.1 | - | 3, 4 | 486 | |||
AK144394.1 | - | 3, 4 | 514 | |||
AK154475.1 | - | 3, 4 | 535 | |||
BC024704.1 | - | 3, 4 | 462 | 4195108 |