Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033146.1 | NP_149158.1 | 4 | 512 | ||
GenBank mRNA | AB054001.1 | - | 4 | 558 | ||
AK005154.1 | - | 4 | 512 | |||
AK144394.1 | - | 4 | 540 | |||
AK154475.1 | - | 4 | 561 | |||
AK180014.1 | - | 2 | 89 | |||
BC024704.1 | - | 4 | 488 | 4195108 |