Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175290.4 | NP_780499.3 | 6 | 2376 | ||
GenBank mRNA | AK049352.1 | - | 4 | 779 | ||
AY596201.1 | - | 7 | 2486 | |||
BC098479.1 | - | 4 | 798 |