Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133832.3 | NP_598593.1 | 5 | 1015 | ||
GenBank mRNA | AF456767.1 | - | 5 | 1123 | ||
AK014244.1 | - | 1 | 683 | |||
AK149656.1 | - | 5 | 1086 | |||
BC019796.1 | - | 5 | 1015 | 4923005 | ||
BC038334.1 | - | 2 | 283 | |||
BC052546.1 | - | 2 | 287 |