Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 9 | 1738 | ||
GenBank mRNA | AK049032.1 | - | 9 | 1742 | ||
AK131145.1 | - | 8 | 3456 | |||
AK157331.1 | - | 9 | 1368 | |||
AK157490.1 | - | 10 | 1160 | |||
AK158435.1 | - | 9 | 1367 | |||
AK165751.1 | - | 9 | 1373 | |||
AK170536.1 | - | 9 | 1766 | |||
AK170847.1 | - | 9 | 1359 | |||
AK170951.1 | - | 9 | 1365 | |||
AK172536.1 | - | 9 | 1365 | |||
BC023470.1 | - | 1 | 70 | |||
BC023752.1 | - | 9 | 1183 | 5354455 | ||
BC031157.1 | - | 9 | 1001 | 4976773 |