Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kti12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029571.2 | NP_083847.1 | 1 | 964 | ||
GenBank mRNA | AK003197.1 | - | 1 | 965 | ||
AK148185.1 | - | 1 | 954 | |||
AK150178.1 | - | 1 | 951 | |||
AK156755.1 | - | 1 | 932 | |||
BC021493.1 | - | 1 | 1047 | 5345710 |