Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 7, 8 | 736 | ||
GenBank mRNA | AB019211.1 | - | 7, 8 | 679 | ||
AK076390.1 | - | 719 | ||||
AK077678.1 | - | 7, 8 | 737 | |||
AK078948.1 | - | 7, 8 | 696 | |||
AK146544.1 | - | 7, 8 | 741 | |||
AK146735.1 | - | 7, 8 | 728 | |||
AK167087.1 | - | 7, 8 | 650 | |||
AK167804.1 | - | 7, 8 | 700 | |||
AK168753.1 | - | 7, 8 | 695 | |||
BC010669.1 | - | 7, 8 | 723 | 2645010 |