Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 9 | 892 | ||
GenBank mRNA | AB019211.1 | - | 9 | 835 | ||
AK076390.1 | - | 875 | ||||
AK077678.1 | - | 9 | 893 | |||
AK078948.1 | - | 9 | 852 | |||
AK146544.1 | - | 9 | 897 | |||
AK146735.1 | - | 9 | 884 | |||
AK167087.1 | - | 9 | 806 | |||
AK167804.1 | - | 9 | 856 | |||
AK168753.1 | - | 9 | 851 | |||
AK170203.1 | - | 9 | 929 | |||
BC010669.1 | - | 9 | 879 | 2645010 |