Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igflr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145580.2 | NP_663555.2 | 4 | 474 | ||
GenBank mRNA | AK154129.1 | - | 4 | 475 | ||
BC019733.1 | - | 3 | 515 | 3676256 |