Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133983.4 | NP_598744.1 | 5 | 1082 | ||
GenBank mRNA | AK031354.1 | - | 5 | 813 | ||
AK155114.1 | - | 6 | 1168 | |||
AY190318.1 | - | 5 | 897 | |||
BC019436.1 | - | 5 | 986 | 3674228 | ||
BC056608.1 | - | 5 | 1083 | 6397680 | ||
DQ832276.1 | - | 6 | 1181 |