Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133983.4 | NP_598744.1 | 2 | 596 | ||
GenBank mRNA | AK031354.1 | - | 2 | 327 | ||
AK155114.1 | - | 3 | 682 | |||
AY190318.1 | - | 2 | 411 | |||
BC019436.1 | - | 2 | 500 | 3674228 | ||
BC056608.1 | - | 2 | 597 | 6397680 | ||
DQ832276.1 | - | 3 | 695 |