Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172099.1 | NP_001165570.1 | 2 | 700 | ||
NM_198013.3 | NP_932130.2 | 3 | 898 | |||
GenBank mRNA | AK049250.1 | - | 3 | 862 | ||
AK162604.1 | - | 2 | 280 | |||
BC024476.1 | - | 3 | 747 | 4976954 | ||
BC025434.1 | - | 3 | 752 | |||
BC029708.1 | - | 3 | 831 | 5041450 | ||
BC131989.1 | - | 2 | 671 | 40130266 | ||
BC131991.1 | - | 2 | 671 | 40130268 | ||
BC145206.1 | - | 2 | 671 | 9053757 | ||
CK332869.1 | - | 2 | 144 |