Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291047.1 | NP_001277976.1 | 5 | 742 | ||
NM_001291053.1 | NP_001277982.1 | 3 | 336 | |||
NM_023249.6 | NP_075738.1 | 4 | 616 | |||
GenBank mRNA | AB098740.1 | - | 3 | 228 | ||
AF190624.1 | - | 4 | 532 | |||
AK005955.1 | - | 4 | 452 | |||
AK006049.1 | - | 4 | 452 | |||
AK006137.1 | - | 3 | 234 | |||
AK006505.1 | - | 4 | 427 | |||
AK006858.1 | - | 4 | 504 | |||
AK011452.1 | - | 4 | 547 | |||
AK014931.1 | - | 4 | 442 | |||
AK015752.1 | - | 3 | 453 | |||
AK018910.1 | - | 4 | 427 | |||
AK019819.1 | - | 4 | 447 | |||
AK076063.1 | - | 4 | 559 | |||
AK133401.1 | - | 4 | 530 | |||
AK139453.1 | - | 4 | 524 | |||
BB649357.1 | - | 4 | 616 | |||
BC049737.1 | - | 3 | 246 | 6771362 | ||
BQ573264.1 | - | 3 | 302 |