Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026924.3 | NP_081200.2 | 3 | 557 | ||
NM_152947.2 | NP_694455.2 | 3 | 298 | |||
GenBank mRNA | AB101294.1 | - | 3 | 460 | ||
AB101295.1 | - | 2 | 502 | |||
AB101296.1 | - | 3 | 632 | |||
AK007360.1 | - | 2 | 183 | |||
AK167697.1 | - | 3 | 548 | |||
AY090537.1 | - | 3 | 368 | |||
AY090538.1 | - | 3 | 298 | |||
BC031771.1 | - | 3 | 557 | 4219296 | ||
BC057210.1 | - | 3 | 578 | 5329625 | ||
BC094445.1 | - | 3 | 569 | 4981833 | ||
BY098454.1 | - | 3 | 298 |