Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175399.4 | NP_780608.1 | 3 | 581 | ||
GenBank mRNA | AK005101.2 | - | 3 | 518 | ||
AK083308.1 | - | 3 | 540 | |||
AK089890.1 | - | 3 | 582 | |||
AK144062.1 | - | 3 | 496 | |||
AK167629.1 | - | 3 | 544 | |||
AK196133.1 | - | 1 | 114 | |||
BC012277.1 | - | 3 | 526 | 4007125 |