Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133823.4 | NP_598584.2 | 3 | 599 | ||
GenBank mRNA | AK050255.1 | - | 3 | 581 | ||
AK160235.1 | - | 4 | 925 | |||
BC021954.1 | - | 2 | 550 | 5134853 | ||
BC085229.1 | - | 2 | 558 |