Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133823.4 | NP_598584.2 | 5 | 851 | ||
GenBank mRNA | AK050255.1 | - | 5 | 833 | ||
AK160235.1 | - | 6 | 1177 | |||
BC021954.1 | - | 4 | 802 | 5134853 | ||
BC027398.1 | - | 3 | 262 | |||
BC085229.1 | - | 4 | 810 |