Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144526.3 | NP_653109.2 | 4, 5 | 737 | ||
GenBank mRNA | AK011345.1 | - | 4, 5 | 735 | ||
AK049710.1 | - | 4, 5 | 738 | |||
AK078429.1 | - | 4, 5 | 738 | |||
AK186477.1 | - | 1, 2 | 72 | |||
BC020115.1 | - | 4, 5 | 723 | 3988382 | ||
BC029115.1 | - | 4, 5 | 723 | 3983148 |