Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145875.1 | NP_001139347.1 | 6 | 857 | ||
NM_175417.4 | NP_780626.1 | 6 | 778 | |||
GenBank mRNA | AK029025.1 | - | 6 | 780 | ||
AK079178.1 | - | 6 | 859 | |||
AK135327.1 | - | 6 | 858 | |||
BC027755.1 | - | 5 | 687 | 5055136 |