Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133768.4 | NP_598529.1 | 17 | 1414 | ||
GenBank mRNA | AK010922.1 | - | 17 | 1426 | ||
AK076453.1 | - | 17 | 1429 | |||
AK138423.1 | - | 17 | 1411 | |||
AK140160.1 | - | 17 | 1429 | |||
AK148764.1 | - | 17 | 1398 | |||
AK168739.1 | - | 17 | 1409 | |||
AK212014.1 | - | 1 | 56 | |||
BC016670.1 | - | 17 | 1414 | 2648753 |