Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290818.1 | NP_001277747.1 | 19 | 1945 | ||
NM_130863.2 | NP_570933.1 | 20 | 2017 | |||
GenBank mRNA | AF333028.1 | - | 19 | 1737 | ||
AK155691.1 | - | 19 | 1946 | |||
BC003196.1 | - | 19 | 1733 | |||
BC033272.1 | - | 16 | 2093 | |||
BC038312.1 | - | 1 | 616 | |||
BC053922.1 | - | 20 | 1750 | 6309555 |