Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007419.2 | NP_031445.2 | 1 | 213 | ||
GenBank mRNA | AK018378.1 | - | 1 | 31 | ||
BC140970.1 | - | 1 | 386 | 9055582 | ||
BC147435.1 | - | 1 | 386 | 9056029 |
Set Membership: |
G-protein coupled receptor |