Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009672.3 | NP_033802.2 | 4 | 544 | ||
GenBank mRNA | AF022957.1 | - | 4 | 453 | ||
AK020753.1 | - | 4 | 506 | |||
BC062899.1 | - | 4 | 459 | 6415168 | ||
U73478.1 | - | 4 | 411 |