Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007457.2 | NP_031483.1 | 3 | 278 | ||
GenBank mRNA | AK002225.1 | - | 3 | 256 | ||
AK213207.1 | - | 2 | 201 | |||
BC052692.1 | - | 3 | 282 | 5714625 | ||
M62418.1 | - | 3 | 278 |