Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109873.1 | NP_001103343.1 | 8, 9 | 1363 | ||
NM_009824.2 | NP_033954.2 | 8, 9 | 1344 | |||
NM_177289.1 | NP_796263.1 | 7, 8 | 1285 | |||
GenBank mRNA | AF038029.1 | - | 8, 9 | 1226 | ||
AK033680.1 | - | 7, 8 | 1181 | |||
AK041887.1 | - | 8, 9 | 1366 | |||
BC141396.1 | - | 8, 9 | 1173 | 9056009 | ||
BC151156.1 | - | 8, 9 | 1173 | 9088044 |