Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 4, 5 | 503 | ||
GenBank mRNA | AF121906.1 | - | 4, 5 | 416 | ||
AF173379.1 | - | 4, 5 | 476 | |||
AK010264.1 | - | 1, 2 | 112 | |||
AK050072.1 | - | 4, 5 | 452 | |||
BC026938.1 | - | 4, 5 | 470 | 4024140 |